Assay Details
Target Gene Details
Entrez Gene ID: | 8924 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase 2 |
Gene Aliases: |
D15F37S1, MRT38, SHEP1, jdf2, p528 |
Location: |
Chr.15:28111037-28322173 on Build GRCh38 |
Assay Gene Location: | Within Exon 90 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HERC2 | NM_004667.5 | 85 | 13244 | NP_004658.3 |
XM_005268276.4 | 84 | 13374 | XP_005268333.1 | |
XM_006720726.3 | 85 | 13250 | XP_006720789.1 | |
XM_006720727.3 | 83 | 13007 | XP_006720790.1 | |
XM_011522131.2 | 83 | 13031 | XP_011520433.1 | |
XM_011522133.2 | 66 | 10010 | XP_011520435.1 | |
XM_017022695.1 | 84 | 13191 | XP_016878184.1 | |
XM_017022696.1 | 84 | 13134 | XP_016878185.1 | |
XM_017022697.1 | 44 | 6573 | XP_016878186.1 | |
XM_017022698.1 | 44 | 6484 | XP_016878187.1 | |
AF071172.3 | 85 | 13196 | AAD08657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635988 | Chr.15:28098445 - 28194723 on Build GRCh38 | Gain |
![]() ![]() |
nsv428299 | Chr.15:28042424 - 28817373 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv568663 | Chr.15:27783102 - 28176880 on Build GRCh38 | Loss |
![]() ![]() |
nsv1039684 | Chr.15:28080438 - 28315658 on Build GRCh38 | Gain |
![]() ![]() |
nsv1052131 | Chr.15:27932591 - 28460005 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1038472 | Chr.15:27647423 - 28256473 on Build GRCh38 | Gain |
![]() ![]() |
nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1047941 | Chr.15:28110984 - 28168961 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
