Assay Details
Target Gene Details
Entrez Gene ID: | 10102 |
Gene Name: | Ts translation elongation factor, mitochondrial |
Gene Aliases: |
EFTS, EFTSMT |
Location: |
Chr.12:57782745-57802856 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSFM | NM_001172695.1 | NP_001166166.1 | ||
| NM_001172696.1 | NP_001166167.1 | |||
| NM_001172697.1 | NP_001166168.1 | |||
| NM_005726.5 | NP_005717.3 | |||
| AF110399.1 | AAD20224.1 | |||
| AK295323.1 | ||||
| AK304621.1 | ||||
| AK308981.1 | ||||
| AK313453.1 | ||||
| BC022862.1 | AAH22862.1 | |||
| BC093068.1 | AAH93068.1 | |||
| L37936.1 | AAC37577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
|
| nsv470297 | Chr.12:57739473 - 57808636 on Build GRCh38 | Gain |
|
| nsv510601 | Chr.12:57736515 - 57798142 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map