Assay Details
Target Gene Details
Entrez Gene ID: | 23098 |
Gene Name: | sterile alpha and TIR motif containing 1 |
Gene Aliases: |
MyD88-5, SAMD2, SARM |
Location: |
Chr.17:28371662-28401045 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARM1 | NM_015077.3 | 9 | 3197 | NP_055892.2 |
| AB011096.1 | 9 | 2449 | BAA25450.1 | |
| AJ290445.1 | 9 | 2752 | CAB90355.1 | |
| AK026397.1 | 1 | 808 | ||
| AK093615.1 | 6 | 1693 | ||
| AK302038.1 | ||||
| BC040429.1 | 9 | 2785 |
Target Gene Details
Entrez Gene ID: | 113235 |
Gene Name: | solute carrier family 46 member 1 |
Gene Aliases: |
G21, HCP1, PCFT |
Location: |
Chr.17:28394642-28406630 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC46A1 | NM_001242366.2 | 4 | 4113 | NP_001229295.1 |
| NM_080669.5 | 5 | 4197 | NP_542400.2 | |
| XM_005277786.3 | 3 | 3956 | XP_005277843.1 | |
| XM_017024110.1 | 6 | 4156 | XP_016879599.1 | |
| AK074161.1 | 3 | 3211 | BAB84987.1 | |
| AL832613.2 | 4 | 3505 | CAD89945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062056 | Chr.17:28376191 - 28452023 on Build GRCh38 | Gain |
|
| nsv1056839 | Chr.17:28348199 - 28428188 on Build GRCh38 | Gain |
|
| nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map