Assay Details
Target Gene Details
Entrez Gene ID: | 4332 |
Gene Name: | myeloid cell nuclear differentiation antigen |
Gene Aliases: |
PYHIN3 |
Location: |
Chr.1:158831378-158849480 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MNDA | NM_002432.1 | NP_002423.1 | ||
| AK290392.1 | ||||
| AK313210.1 | ||||
| BC020715.1 | ||||
| BC032319.1 | AAH32319.1 | |||
| M81750.1 | AAA69696.1 |
Target Gene Details
Entrez Gene ID: | 128372 |
Gene Name: | olfactory receptor family 6 subfamily N member 1 |
Gene Aliases: |
OR1-22 |
Location: |
Chr.1:158747733-158840979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6N1 | XM_017000325.1 | XP_016855814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv589n54 | Chr.1:158715246 - 158883163 on Build GRCh38 | Loss |
|
| nsv1002401 | Chr.1:158742837 - 159002241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map