Assay Details
Target Gene Details
Entrez Gene ID: | 6863 |
Gene Name: | tachykinin precursor 1 |
Gene Aliases: |
Hs.2563, NK2, NKNA, NPK, TAC2 |
Location: |
Chr.7:97731959-97740472 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAC1 | NM_003182.2 | 1 | 120 | NP_003173.1 |
| NM_013996.2 | 1 | 120 | NP_054702.1 | |
| NM_013997.2 | 1 | 120 | NP_054703.1 | |
| NM_013998.2 | 1 | 120 | NP_054704.1 | |
| AK222957.1 | BAD96677.1 | |||
| BC018047.1 | AAH18047.1 | |||
| DN992454.1 | 1 | 128 | ||
| U37529.1 | AAA79195.1 | |||
| X54469.1 | CAA38351.1 |
More Information
Set Membership: |
|

Genomic Map