Assay Details
Target Gene Details
Entrez Gene ID: | 1185 |
Gene Name: | chloride voltage-gated channel 6 |
Gene Aliases: |
CLC-6 |
Location: |
Chr.1:11806096-11843144 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN6 | NM_001256959.1 | NP_001243888.1 | ||
| NM_001286.3 | NP_001277.1 | |||
| NR_046428.1 | ||||
| DA229615.1 |
Target Gene Details
Entrez Gene ID: | 4524 |
Gene Name: | methylenetetrahydrofolate reductase (NAD(P)H) |
Gene Aliases: |
- |
Location: |
Chr.1:11785730-11806103 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFR | NM_005957.4 | NP_005948.3 | ||
| XM_005263460.4 | XP_005263517.1 | |||
| XM_005263463.3 | XP_005263520.1 | |||
| AY046560.1 | 1 | 842 | AAL17646.1 |
More Information
Set Membership: |
|

Genomic Map