Assay Details
Target Gene Details
Entrez Gene ID: | 57119 |
Gene Name: | epididymal peptidase inhibitor |
Gene Aliases: |
CT71, CT72, SPINLW1, WAP7, WFDC7, dJ461P17.2 |
Location: |
Chr.20:45540626-45547426 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPPIN | NM_001302861.1 | 4 | 616 | NP_001289790.1 |
| NM_020398.3 | 4 | 605 | NP_065131.1 | |
| AF286368.1 | 4 | 557 | AAG00546.1 | |
| AF286369.1 | 3 | 1052 | AAG00547.1 | |
| AF286370.2 | 4 | 557 | AAG00548.1 | |
| AK126022.1 | 2 | 3852 | ||
| AL118493.1 | 4 | 559 | CAB56343.1 | |
| BC044829.1 | 4 | 568 | ||
| BC053369.1 | 4 | 563 | AAH53369.1 | |
| BF057305.1 | 1 | 169 | ||
| DA821702.1 | ||||
| DC395362.1 |
Target Gene Details
Entrez Gene ID: | 100526773 |
Gene Name: | EPPIN-WFDC6 readthrough |
Gene Aliases: |
SPINLW1-WFDC6 |
Location: |
Chr.20:45536280-45547426 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPPIN-WFDC6 | NM_001198986.1 | NP_001185915.1 | ||
| AK301937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map