Assay Details
Target Gene Details
Entrez Gene ID: | 5226 |
Gene Name: | phosphogluconate dehydrogenase |
Gene Aliases: |
6PGD |
Location: |
Chr.1:10398992-10420511 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGD | NM_001304451.1 | NP_001291380.1 | ||
| NM_001304452.1 | NP_001291381.1 | |||
| NM_002631.3 | NP_002622.2 | |||
| AK290404.1 | ||||
| AK296889.1 | ||||
| AK298830.1 | ||||
| AK300952.1 | ||||
| AK303898.1 | ||||
| AK304423.1 | ||||
| BC000368.2 | AAH00368.1 | |||
| U30255.1 | AAA75302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1010529 | Chr.1:10416138 - 10418529 on Build GRCh38 | Deletion |
|
| esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain |
|
| esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map