Assay Details
Target Gene Details
Entrez Gene ID: | 1982 |
Gene Name: | eukaryotic translation initiation factor 4 gamma 2 |
Gene Aliases: |
AAG1, DAP5, NAT1, P97 |
Location: |
Chr.11:10797046-10809035 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4G2 | NM_001042559.2 | NP_001036024.3 | ||
| NM_001172705.1 | NP_001166176.1 | |||
| NM_001418.3 | NP_001409.3 | |||
| AB209267.1 | BAD92504.1 | |||
| AK123338.1 | ||||
| AK223548.1 | BAD97268.1 | |||
| AK302886.1 | ||||
| AK310387.1 | ||||
| AY513274.1 | AAT08027.1 | |||
| BC018746.1 | ||||
| BC018975.2 | ||||
| BC039851.1 | AAH39851.1 | |||
| BC043149.1 | ||||
| BC065276.1 | ||||
| BC111415.1 | ||||
| BC111548.1 | ||||
| BX647799.1 | ||||
| U73824.1 | AAB49973.1 | |||
| U76111.1 | AAC51166.1 | |||
| X89713.1 | CAA61857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map