Assay Details
Target Gene Details
Entrez Gene ID: | 3418 |
Gene Name: | isocitrate dehydrogenase (NADP(+)) 2, mitochondrial |
Gene Aliases: |
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH |
Location: |
Chr.15:90083978-90102554 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH2 | NM_001289910.1 | 7 | 815 | NP_001276839.1 |
| NM_001290114.1 | 5 | 790 | NP_001277043.1 | |
| NM_002168.3 | 7 | 1048 | NP_002159.2 | |
| AK222916.1 | 7 | 962 | BAD96636.1 | |
| AK294148.1 | 7 | 815 | ||
| AK299987.1 | 5 | 704 | ||
| AK312627.1 | 7 | 1012 | ||
| AK316388.1 | 7 | 889 | ||
| BC009244.2 | 7 | 950 | AAH09244.1 | |
| BC071828.1 | 7 | 949 | AAH71828.1 | |
| X69433.1 | 7 | 970 | CAA49208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637213 | Chr.15:90084539 - 90105288 on Build GRCh38 | Gain |
|
| dgv112e55 | Chr.15:90079272 - 90249274 on Build GRCh38 | Gain |
|
| esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain |
|
| esv3569864 | Chr.15:90056527 - 90089206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map