Assay Details
Target Gene Details
Entrez Gene ID: | 196410 |
Gene Name: | methyltransferase like 7B |
Gene Aliases: |
ALDI |
Location: |
Chr.12:55681546-55684611 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL7B | NM_152637.2 | 1 | 222 | NP_689850.2 |
| AK290112.1 | 1 | 222 | ||
| AY358508.1 | 1 | 90 | AAQ88872.1 |
More Information
Set Membership: |
|

Genomic Map