Assay Details
Target Gene Details
| Entrez Gene ID: | 83706 | 
| Gene Name: | fermitin family member 3 | 
| Gene Aliases: | KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF | 
| Location: | Chr.11:64206665-64223891 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 3 - Exon 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| FERMT3 | NM_031471.5 | NP_113659.3 | ||
| NM_178443.2 | NP_848537.1 | |||
| XM_011545294.2 | XP_011543596.1 | |||
| XM_011545295.2 | XP_011543597.1 | |||
| XM_017018398.1 | XP_016873887.1 | |||
| XM_017018399.1 | XP_016873888.1 | |||
| AK093719.1 | BAC04220.1 | |||
| AL560215.3 | ||||
| AY093951.1 | AAM19736.1 | |||
| AY093952.1 | AAM19737.1 | |||
| BC004347.1 | AAH04347.2 | |||
| BC013366.2 | AAH13366.1 | |||
| BC015584.2 | AAH15584.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion |  MACROD1  LOC105369340  BAD  TRPT1  FERMT3  DNAJC4  FKBP2  GPR137  PPP1R14B  NUDT22  STIP1  PLCB3  VEGFB | 
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |  MACROD1  LOC105369340  COX8A  BAD  TRPT1  KCNK4  FKBP2  NAA40  OTUB1  TRMT112  PPP1R14B  PRDX5  PLCB3  CCDC88B  ESRRA  FLRT1  FERMT3  MIR7155  DNAJC4  TEX40  KCNK4-TEX40  GPR137  NUDT22  STIP1  VEGFB | 
| dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |  ESRRA  LOC105369340  BAD  TRPT1  FERMT3  KCNK4  DNAJC4  FKBP2  TEX40  TRMT112  KCNK4-TEX40  GPR137  PPP1R14B  NUDT22  STIP1  PRDX5  PLCB3  VEGFB | 
| nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss |  PPP1R14B  LOC105369340  NUDT22  TRPT1  STIP1  FERMT3  PLCB3  DNAJC4  VEGFB  FKBP2 | 
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |  ESRRA  LOC105369340  BAD  TRPT1  FERMT3  KCNK4  MIR7155  DNAJC4  FKBP2  TEX40  TRMT112  KCNK4-TEX40  GPR137  PPP1R14B  MIR1237  NUDT22  PRDX5  PLCB3  CCDC88B  RPS6KA4  VEGFB | 
| nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss |  MACROD1  LOC105369340  BAD  TRPT1  FERMT3  DNAJC4  FKBP2  GPR137  PPP1R14B  NUDT22  STIP1  PLCB3  VEGFB | 
| esv33774 | Chr.11:64210622 - 64211272 on Build GRCh38 | Gain |  FERMT3 | 
| nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss |  MACROD1  ESRRA  LOC105369340  BAD  TRPT1  FLRT1  FERMT3  KCNK4  DNAJC4  FKBP2  TEX40  TRMT112  KCNK4-TEX40  GPR137  PPP1R14B  NUDT22  STIP1  PRDX5  PLCB3  VEGFB | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top