Assay Details
Target Gene Details
Entrez Gene ID: | 6560 |
Gene Name: | solute carrier family 12 member 4 |
Gene Aliases: |
CTC-479C5.17, KCC1, hKCC1 |
Location: |
Chr.16:67943474-67968694 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A4 | NM_001145961.1 | 24 | 3401 | NP_001139433.1 |
| NM_001145962.1 | 23 | 3324 | NP_001139434.1 | |
| NM_001145963.1 | 24 | 3298 | NP_001139435.1 | |
| NM_001145964.1 | 24 | 3289 | NP_001139436.1 | |
| NM_005072.4 | 24 | 3419 | NP_005063.1 | |
| AF047338.1 | 24 | 3278 | AAC32815.1 | |
| AF054505.1 | 23 | 3426 | AAC39684.1 | |
| AF054506.1 | 23 | 3433 | AAC39685.1 | |
| AK026911.1 | 16 | 2057 | ||
| AK097808.1 | ||||
| AK293906.1 | 24 | 3310 | ||
| AK293956.1 | 23 | 3214 | ||
| AK299042.1 | 24 | 3205 | ||
| AK302790.1 | 24 | 3298 | ||
| AK316415.1 | 24 | 3279 | ||
| BC021193.2 | 24 | 3347 | AAH21193.1 | |
| CF272417.1 | 2 | 140 | ||
| U55054.1 | 24 | 3333 | AAC50563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map