Assay Details
Target Gene Details
Entrez Gene ID: | 113220 |
Gene Name: | kinesin family member 12 |
Gene Aliases: |
- |
Location: |
Chr.9:114091623-114100099 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF12 | NM_138424.1 | 16 | 1699 | NP_612433.1 |
| XM_005251683.4 | 20 | 2708 | XP_005251740.1 | |
| XM_006716947.2 | 19 | 1960 | XP_006717010.1 | |
| XM_011518190.2 | 20 | 2743 | XP_011516492.1 | |
| XM_011518191.2 | 19 | 2722 | XP_011516493.1 | |
| XM_017014240.1 | 14 | 1399 | XP_016869729.1 | |
| BC010626.1 | 16 | 1699 | AAH10626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761285 | Chr.9:114049617 - 114340988 on Build GRCh38 | Gain |
|
| nsv528822 | Chr.9:114053497 - 114104774 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| esv3621549 | Chr.9:113836116 - 114331078 on Build GRCh38 | Gain |
|
| nsv515856 | Chr.9:114091517 - 114104774 on Build GRCh38 | Gain+Loss |
|
| dgv945n27 | Chr.9:114048610 - 114104774 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117751044] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map