Assay Details
Target Gene Details
Entrez Gene ID: | 151126 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
ZNF533 |
Location: |
Chr.2:179441982-179864550 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF385B | NM_001113397.1 | 6 | 1069 | NP_001106868.1 |
NM_001113398.1 | 6 | 929 | NP_001106869.1 | |
NM_001282725.1 | 8 | 1023 | NP_001269654.1 | |
NM_152520.4 | 8 | 1596 | NP_689733.3 | |
NR_104234.1 | 10 | 1267 | ||
XM_011510713.2 | 10 | 4737 | XP_011509015.1 | |
XM_011510714.2 | 9 | 4687 | XP_011509016.1 | |
XM_011510715.2 | 9 | 4640 | XP_011509017.1 | |
XM_011510716.2 | 9 | 1848 | XP_011509018.1 | |
XM_011510717.2 | 9 | 1845 | XP_011509019.1 | |
XM_011510719.2 | 9 | 1108 | XP_011509021.1 | |
XM_011510720.2 | 8 | 968 | XP_011509022.1 | |
XM_011510721.2 | 7 | 3261 | XP_011509023.1 | |
XM_017003435.1 | 8 | 1277 | XP_016858924.1 | |
XM_017003436.1 | 8 | 1755 | XP_016858925.1 | |
XM_017003437.1 | 7 | 912 | XP_016858926.1 | |
XM_017003438.1 | 6 | 6395 | XP_016858927.1 | |
XM_017003439.1 | 6 | 903 | XP_016858928.1 | |
XM_017003440.1 | 7 | 4432 | XP_016858929.1 | |
AK057999.1 | 6 | 929 | BAB71629.1 | |
AK096822.1 | 6 | 1069 | BAC04870.1 | |
BC038422.1 | 8 | 1021 | AAH38422.1 | |
BC048123.1 | 10 | 1270 | AAH48123.1 | |
BC092423.1 | 8 | 1431 | AAH92423.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
