Assay Details
Target Gene Details
Entrez Gene ID: | 55347 |
Gene Name: | abhydrolase domain containing 10 |
Gene Aliases: |
- |
Location: |
Chr.3:111978876-111993368 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD10 | NM_001272069.1 | 6 | 1059 | NP_001258998.1 |
| NM_018394.3 | 5 | 1007 | NP_060864.1 | |
| NR_073570.1 | 4 | 823 | ||
| NR_073571.1 | 3 | 711 | ||
| AK002204.1 | 5 | 831 | BAA92133.1 | |
| AK300018.1 | 4 | 689 | ||
| AL713726.1 | 4 | 605 | CAD28516.2 | |
| BC014516.2 | 5 | 846 | AAH14516.1 | |
| DB494690.1 |
More Information
Set Membership: |
|

Genomic Map