Assay Details
Target Gene Details
Entrez Gene ID: | 10419 |
Gene Name: | protein arginine methyltransferase 5 |
Gene Aliases: |
HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs |
Location: |
Chr.14:22920511-22929585 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT5 | NM_001039619.2 | NP_001034708.1 | ||
| NM_001282953.1 | NP_001269882.1 | |||
| NM_001282954.1 | NP_001269883.1 | |||
| NM_001282955.1 | NP_001269884.1 | |||
| NM_001282956.1 | NP_001269885.1 | |||
| NM_006109.4 | NP_006100.2 | |||
| XM_017020927.1 | XP_016876416.1 | |||
| AA496357.1 | ||||
| AF167572.1 | AAF04502.1 | |||
| AK075251.1 | ||||
| AK301812.1 | ||||
| AL833612.1 | ||||
| BC025979.2 | AAH25979.1 |
Target Gene Details
Entrez Gene ID: | 100505758 |
Gene Name: | PRMT5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:22919456-22923407 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT5-AS1 | NR_120599.1 | 1 | 1079 | |
| NR_120600.1 | 1 | 1079 | ||
| AK093277.1 | 1 | 1079 |
More Information
Set Membership: |
|

Genomic Map