Assay Details
Target Gene Details
| Entrez Gene ID: | 404216 | 
| Gene Name: | long intergenic non-protein coding RNA 1561 | 
| Gene Aliases: | C10orf85 | 
| Location: | Chr.10:120597949-120600123 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LINC01561 | NR_103717.1 | 1 | 436 | |
| AK094721.1 | 1 | 176 | BAC04408.1 | |
| BC108662.1 | 1 | 436 | AAI08663.1 | 
Target Gene Details
| Entrez Gene ID: | 196051 | 
| Gene Name: | phospholipid phosphatase 4 | 
| Gene Aliases: | DPPL2, PPAPDC1, PPAPDC1A | 
| Location: | Chr.10:120456954-120600123 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 11 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PLPP4 | XM_005269592.2 | 6 | 9879 | XP_005269649.1 | 
| XM_006717685.3 | 7 | 9719 | XP_006717748.1 | |
| XM_011539444.2 | 8 | 9777 | XP_011537746.1 | |
| XM_017015821.1 | 4 | 9460 | XP_016871310.1 | |
| XM_017015822.1 | 4 | 9427 | XP_016871311.1 | |
| DR000933.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3624731 | Chr.10:120590572 - 120713656 on Build GRCh38 | Gain |  LINC01561  PLPP4 | 
| nsv528787 | Chr.10:120586553 - 120722450 on Build GRCh38 | Gain |  LINC01561  PLPP4 | 
| esv2760128 | Chr.10:120590025 - 120720150 on Build GRCh38 | Gain |  LINC01561  PLPP4 | 
| nsv1052357 | Chr.10:120592928 - 120710371 on Build GRCh38 | Gain |  LINC01561  PLPP4 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top