Assay Details
Target Gene Details
Entrez Gene ID: | 50810 |
Gene Name: | hepatoma-derived growth factor, related protein 3 |
Gene Aliases: |
CGI-142, HDGF-2, HDGF2, HRP-3 |
Location: |
Chr.15:83112738-83208018 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDGFRP3 | XM_006720554.3 | XP_006720617.1 |
Target Gene Details
Entrez Gene ID: | 53346 |
Gene Name: | transmembrane 6 superfamily member 1 |
Gene Aliases: |
- |
Location: |
Chr.15:83107572-83145403 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
More Information
Set Membership: |
|

Genomic Map