Assay Details
Target Gene Details
Entrez Gene ID: | 23761 |
Gene Name: | phosphatidylserine decarboxylase |
Gene Aliases: |
DJ858B16, PSD, PSDC, PSSC, dJ858B16.2 |
Location: |
Chr.22:31618491-31630824 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PISD | NM_001326411.1 | 4 | 830 | NP_001313340.1 |
| NM_001326412.1 | 4 | 767 | NP_001313341.1 | |
| NM_001326413.1 | 4 | 916 | NP_001313342.1 | |
| NM_001326414.1 | 5 | 996 | NP_001313343.1 | |
| NM_001326415.1 | 5 | 1170 | NP_001313344.1 | |
| NM_001326416.1 | 7 | 1359 | NP_001313345.1 | |
| NM_001326417.1 | 6 | 1250 | NP_001313346.1 | |
| NM_001326418.1 | 3 | 750 | NP_001313347.1 | |
| NM_001326419.1 | 6 | 1250 | NP_001313348.1 | |
| NM_001326420.1 | 3 | 750 | NP_001313349.1 | |
| NM_001326421.1 | 4 | 830 | NP_001313350.1 | |
| NM_014338.3 | 5 | 802 | NP_055153.1 | |
| NM_178022.1 | 4 | 711 | NP_821141.1 | |
| AK298474.1 | 4 | 711 | ||
| AL050371.1 | 4 | 481 | CAB43678.2 | |
| BC001482.2 | 5 | 802 | AAH01482.1 | |
| CR456540.1 | 4 | 480 | CAG30426.1 | |
| DQ655940.2 | ||||
| DQ656048.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829176 | Chr.22:31602237 - 31633570 on Build GRCh38 | Loss |
|
| nsv517534 | Chr.22:31581608 - 31628994 on Build GRCh38 | Loss |
|
| nsv953070 | Chr.22:31609915 - 31630414 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map