Assay Details
Target Gene Details
Entrez Gene ID: | 1876 |
Gene Name: | E2F transcription factor 6 |
Gene Aliases: |
E2F-6 |
Location: |
Chr.2:11444375-11466177 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Exon 9 |
More Information
Set Membership: |
|

Genomic Map