Assay Details
Target Gene Details
Entrez Gene ID: | 2829 |
Gene Name: | X-C motif chemokine receptor 1 |
Gene Aliases: |
CCXCR1, GPR5 |
Location: |
Chr.3:46016990-46086803 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XCR1 | NM_001024644.1 | NP_001019815.1 | ||
| NM_005283.2 | NP_005274.1 | |||
| XM_017006196.1 | 4 | 2340 | XP_016861685.1 | |
| XM_017006197.1 | 6 | 2592 | XP_016861686.1 | |
| XM_017006198.1 | 5 | 2405 | XP_016861687.1 | |
| XM_017006199.1 | 4 | 1402 | XP_016861688.1 | |
| XM_017006200.1 | 3 | 2260 | XP_016861689.1 | |
| XM_017006201.1 | 2 | 2109 | XP_016861690.1 | |
| BC095533.1 | AAH95533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759145 | Chr.3:45974972 - 46078908 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map