Assay Details
Target Gene Details
Entrez Gene ID: | 404220 |
Gene Name: | chromosome 6 open reading frame 201 |
Gene Aliases: |
dJ1013A10.5 |
Location: |
Chr.6:4079197-4130788 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf201 | NM_001085401.2 | 3 | 901 | NP_001078870.1 |
NR_104463.1 | 3 | 901 | ||
NR_104464.1 | 2 | 523 | ||
XM_005249112.3 | 2 | 378 | XP_005249169.1 | |
XM_011514613.2 | 3 | 515 | XP_011512915.1 | |
XM_017010861.1 | 2 | 532 | XP_016866350.1 | |
AF429972.1 | 2 | 351 | AAP97304.1 | |
AK124114.1 | 2 | 336 | ||
AK301893.1 | 2 | 336 | ||
BC047663.1 | 3 | 898 | AAH47663.1 |
