Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3 | NM_001012338.2 | 20 | 2749 | NP_001012338.1 |
| NM_001243101.1 | 18 | 2683 | NP_001230030.1 | |
| NM_002530.3 | 19 | 2707 | NP_002521.2 | |
| XM_006720543.3 | 17 | 3016 | XP_006720606.1 | |
| XM_006720544.3 | 16 | 2992 | XP_006720607.1 | |
| XM_017022240.1 | 18 | 2887 | XP_016877729.1 | |
| XM_017022241.1 | 17 | 2372 | XP_016877730.1 | |
| XM_017022243.1 | 17 | 2368 | XP_016877732.1 | |
| XM_017022244.1 | 17 | 2270 | XP_016877733.1 | |
| XM_017022245.1 | 18 | 2325 | XP_016877734.1 | |
| XM_017022246.1 | 17 | 2426 | XP_016877735.1 | |
| XM_017022251.1 | 10 | 1387 | XP_016877740.1 | |
| XM_017022252.1 | 9 | 1363 | XP_016877741.1 | |
| XM_017022253.1 | 18 | 3058 | XP_016877742.1 | |
| XM_017022254.1 | 18 | 3091 | XP_016877743.1 | |
| AF052184.1 | 8 | 1169 | ||
| AI613045.1 | ||||
| AK297160.1 | 16 | 2401 | ||
| AK313646.1 | 16 | 2402 | ||
| KJ534904.1 | 18 | 2443 | ||
| KJ534905.1 | 17 | 2401 | ||
| KJ534906.1 | 16 | 2377 | ||
| KJ535052.1 | 17 | 2540 | ||
| S76475.1 | 18 | 2458 | AAB33111.1 | |
| U05012.1 | 18 | 2556 | AAA75374.1 |
More Information
Set Membership: |
|

Genomic Map