Assay Details
Target Gene Details
Entrez Gene ID: | 10419 |
Gene Name: | protein arginine methyltransferase 5 |
Gene Aliases: |
HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs |
Location: |
Chr.14:22920511-22929585 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT5 | NM_001039619.2 | NP_001034708.1 | ||
| NM_001282953.1 | NP_001269882.1 | |||
| NM_001282954.1 | NP_001269883.1 | |||
| NM_001282955.1 | NP_001269884.1 | |||
| NM_001282956.1 | NP_001269885.1 | |||
| NM_006109.4 | NP_006100.2 | |||
| XM_017020927.1 | XP_016876416.1 | |||
| AB451246.1 | ||||
| AB451370.1 | ||||
| AF015913.1 | AAB66581.1 | |||
| AF167572.1 | AAF04502.1 | |||
| AK075251.1 | ||||
| AK300863.1 | ||||
| AK301812.1 | ||||
| AK302240.1 | ||||
| AK315500.1 | ||||
| AL833612.1 | ||||
| BC025979.2 | AAH25979.1 | |||
| CR456741.1 | CAG33022.1 |
More Information
Set Membership: |
|

Genomic Map