Assay Details
Target Gene Details
Entrez Gene ID: | 2109 |
Gene Name: | electron transfer flavoprotein beta subunit |
Gene Aliases: |
FP585, MADD |
Location: |
Chr.19:51345155-51366418 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFB | NM_001014763.1 | 2 | 819 | NP_001014763.1 |
| NM_001985.2 | 3 | 434 | NP_001976.1 | |
| AF370381.1 | 2 | 801 | AAQ15217.1 | |
| AK055285.1 | 2 | 821 | ||
| AK291881.1 | 3 | 361 | ||
| AL833205.1 | 2 | 3095 | ||
| BC093961.1 | 3 | 365 | AAH93961.1 | |
| BC093963.1 | 3 | 364 | AAH93963.1 | |
| BG281641.1 | 2 | 252 | ||
| BG399783.1 | 3 | 435 | ||
| CR456827.1 | 3 | 342 | CAG33108.1 | |
| X71129.1 | 3 | 369 | CAA50441.1 |
More Information
Set Membership: |
|

Genomic Map