Assay Details
Target Gene Details
Entrez Gene ID: | 51559 |
Gene Name: | 5'-nucleotidase domain containing 3 |
Gene Aliases: |
TU12B1-TY, TU12B1TY |
Location: |
Chr.12:103746315-103841238 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5DC3 |
Target Gene Details
Entrez Gene ID: | 55576 |
Gene Name: | stabilin 2 |
Gene Aliases: |
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1 |
Location: |
Chr.12:103587250-103766724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 66 - Exon 67 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAB2 | NM_017564.9 | NP_060034.9 | ||
| XM_011538539.2 | XP_011536841.1 | |||
| XM_011538541.2 | XP_011536843.1 | |||
| XM_011538542.2 | XP_011536844.1 | |||
| AB052958.1 | BAC15608.1 | |||
| AF160476.1 | AAF82398.1 | |||
| AJ295695.1 | CAC82105.1 | |||
| AK024503.1 | BAB15793.1 | |||
| AK074051.1 | BAB84877.1 | |||
| AK160380.1 | BAD18723.1 | |||
| AL133021.1 | CAB61358.2 | |||
| AY227444.1 | AAO39681.1 | |||
| AY311388.1 | AAP74958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34031 | Chr.12:103720499 - 103904768 on Build GRCh38 | Loss |
|
| esv3892262 | Chr.12:103705376 - 103817662 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map