Assay Details
Target Gene Details
Entrez Gene ID: | 60370 |
Gene Name: | arginine vasopressin induced 1 |
Gene Aliases: |
PP5395, VIP32, VIT32 |
Location: |
Chr.10:97677422-97687432 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVPI1 | NM_021732.2 | 2 | 677 | NP_068378.2 |
| XM_017016494.1 | 2 | 837 | XP_016871983.1 | |
| AF131791.1 | 2 | 677 | ||
| AF218019.1 | 2 | 632 | AAG17261.1 | |
| AF241786.1 | 2 | 680 | AAG44474.1 | |
| AK024533.1 | 2 | 649 | ||
| AK222493.1 | 2 | 650 | BAD96213.1 | |
| AK223099.1 | 2 | 625 | BAD96819.1 | |
| BC000877.1 | 2 | 288 | AAH00877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045292 | Chr.10:97651396 - 97691104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map