Assay Details
Target Gene Details
Entrez Gene ID: | 29893 |
Gene Name: | PSMC3 interacting protein |
Gene Aliases: |
GT198, HOP2, HUMGT198A, ODG3, TBPIP |
Location: |
Chr.17:42572310-42577831 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC3IP | NM_001256014.1 | NP_001242943.1 | ||
| NM_001256015.1 | NP_001242944.1 | |||
| NM_001256016.1 | NP_001242945.1 | |||
| NM_013290.6 | NP_037422.2 | |||
| NM_016556.3 | NP_057640.1 | |||
| NR_045669.1 | ||||
| NR_045670.1 | ||||
| NR_045671.1 | ||||
| AB030304.1 | BAA92872.1 | |||
| AF440240.1 | AAL33609.1 | |||
| AK126369.1 | ||||
| BC008792.1 | AAH08792.1 | |||
| BI115923.1 | ||||
| FJ952179.1 | ||||
| FJ952180.1 | ||||
| FJ952181.1 | ||||
| FJ952182.1 | ||||
| FJ952183.1 | ||||
| GQ851964.1 | ||||
| GQ851965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map