Assay Details
Target Gene Details
Entrez Gene ID: | 51479 |
Gene Name: | ankyrin repeat and FYVE domain containing 1 |
Gene Aliases: |
ANKHZN, BTBD23, ZFYVE14 |
Location: |
Chr.17:4163371-4264026 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKFY1 | NM_001257999.1 | 25 | 8093 | NP_001244928.1 |
| NM_016376.3 | 25 | 7891 | NP_057460.3 | |
| NR_047571.1 | 24 | 7772 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065237 | Chr.17:4040089 - 4278208 on Build GRCh38 | Gain |
|
| dgv557e214 | Chr.17:4142251 - 4216890 on Build GRCh38 | Gain |
|
| dgv3086n100 | Chr.17:4101015 - 4199530 on Build GRCh38 | Gain |
|
| nsv457647 | Chr.17:4102227 - 4170104 on Build GRCh38 | Gain |
|
| esv2751682 | Chr.17:4116672 - 4186588 on Build GRCh38 | Gain |
|
| dgv173n21 | Chr.17:4144787 - 4182539 on Build GRCh38 | Gain |
|
| nsv1065665 | Chr.17:4141953 - 4210380 on Build GRCh38 | Gain |
|
| nsv833344 | Chr.17:4015060 - 4205271 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map