Assay Details
Target Gene Details
Entrez Gene ID: | 63924 |
Gene Name: | cell death inducing DFFA like effector c |
Gene Aliases: |
CIDE-3, CIDE3, FPLD5, FSP27 |
Location: |
Chr.3:9866710-9880254 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIDEC | NM_001199551.1 | NP_001186480.1 | ||
| NM_001199552.1 | NP_001186481.1 | |||
| NM_001199623.1 | NP_001186552.1 | |||
| NM_001321142.1 | NP_001308071.1 | |||
| NM_001321143.1 | NP_001308072.1 | |||
| NM_001321144.1 | NP_001308073.1 | |||
| NM_022094.3 | NP_071377.2 | |||
| AF303893.1 | AAN32612.1 | |||
| AK024524.1 | BAB14920.1 | |||
| AK024530.1 | BAB14922.1 | |||
| AL598630.1 | ||||
| AY364638.1 | AAR23106.1 | |||
| AY364640.1 | AAQ65242.1 | |||
| BC016851.1 | AAH16851.1 | |||
| BQ695178.1 | ||||
| CD518729.1 | ||||
| CD518776.1 | ||||
| CD519587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595254 | Chr.3:9866873 - 9879489 on Build GRCh38 | Gain |
|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79419480] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map