Assay Details
Target Gene Details
Entrez Gene ID: | 10207 |
Gene Name: | PATJ, crumbs cell polarity complex component |
Gene Aliases: |
Cipp, INADL, InaD-like, hINADL |
Location: |
Chr.1:61742477-62163920 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PATJ | NM_176877.2 | 6 | 717 | NP_795352.2 |
| XM_005270341.3 | 6 | 715 | XP_005270398.1 | |
| XM_005270347.1 | 6 | 715 | XP_005270404.1 | |
| XM_006710278.3 | 6 | 715 | XP_006710341.1 | |
| XM_011540462.2 | 6 | 715 | XP_011538764.1 | |
| XM_011540463.2 | 6 | 903 | XP_011538765.1 | |
| XM_011540464.2 | 6 | 715 | XP_011538766.1 | |
| XM_011540465.2 | 6 | 715 | XP_011538767.1 | |
| XM_011540466.2 | 6 | 715 | XP_011538768.1 | |
| XM_011540467.2 | 6 | 715 | XP_011538769.1 | |
| XM_011540468.2 | 6 | 715 | XP_011538770.1 | |
| XM_011540469.2 | 6 | 715 | XP_011538771.1 | |
| XM_011540470.1 | 6 | 715 | XP_011538772.1 | |
| XM_016999998.1 | 6 | 715 | XP_016855487.1 | |
| XM_016999999.1 | 6 | 715 | XP_016855488.1 | |
| XM_017000000.1 | XP_016855489.1 | |||
| AF397170.1 | 5 | 603 | AAM28433.1 | |
| AJ001306.1 | 6 | 717 | CAA04666.1 | |
| AJ224747.1 | 6 | 717 | CAA12112.1 | |
| AJ224748.1 | 6 | 717 | CAA12113.1 | |
| AK304355.1 | 1 | 74 | ||
| AK307713.1 | 6 | 714 | ||
| AK308428.1 | 6 | 714 | ||
| BC142661.1 | 6 | 700 | ||
| BC142723.1 | 6 | 700 |
More Information
Set Membership: |
|

Genomic Map