Assay Details
Target Gene Details
Entrez Gene ID: | 72 |
Gene Name: | actin, gamma 2, smooth muscle, enteric |
Gene Aliases: |
ACT, ACTA3, ACTE, ACTL3, ACTSG, VSCM |
Location: |
Chr.2:73892966-73919653 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTG2 | NM_001199893.1 | 2 | 141 | NP_001186822.1 |
| NM_001615.3 | 2 | 141 | NP_001606.1 | |
| AK124338.1 | 3 | 662 | ||
| AK304523.1 | 2 | 99 | ||
| AK310215.1 | 2 | 99 | ||
| AK311011.1 | 2 | 99 | ||
| AK311286.1 | 2 | 101 | ||
| AK312955.1 | 2 | 99 | ||
| BC012617.1 | 2 | 103 | AAH12617.1 | |
| BC094877.1 | 1 | 88 | AAH94877.1 | |
| BI850192.1 | 2 | 141 | ||
| CR536515.1 | CAG38753.1 | |||
| CR541794.1 | CAG46593.1 | |||
| X16940.1 | 2 | 90 | CAA34814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582213 | Chr.2:73893658 - 73902347 on Build GRCh38 | Loss |
|
| nsv1010401 | Chr.2:73901234 - 73923975 on Build GRCh38 | Gain |
|
| esv33667 | Chr.2:73901122 - 73901334 on Build GRCh38 | Gain |
|
| nsv470469 | Chr.2:73895096 - 73910717 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map