Assay Details
Target Gene Details
Entrez Gene ID: | 60681 |
Gene Name: | FK506 binding protein 10 |
Gene Aliases: |
BRKS1, FKBP65, OI11, OI6, PPIASE, hFKBP65 |
Location: |
Chr.17:41812262-41823217 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP10 | NM_021939.3 | 10 | 2188 | NP_068758.3 |
| XM_011525099.2 | 11 | 2693 | XP_011523401.1 | |
| XM_011525100.2 | 10 | 1820 | XP_011523402.1 | |
| AB045981.1 | 10 | 1949 | BAB20974.1 | |
| AK000690.1 | ||||
| AK025694.1 | 9 | 1620 | BAB15220.1 | |
| AK025874.1 | 7 | 1237 | BAB15266.1 | |
| AK074413.1 | ||||
| AK092042.1 | 9 | 2137 | ||
| AK092708.1 | 9 | 1852 | BAC03954.1 | |
| AK172811.1 | 10 | 1894 | BAD18781.1 | |
| AK296107.1 | 10 | 1685 | ||
| AL365371.1 | ||||
| AL832985.1 | 10 | 1930 | CAH56332.1 | |
| BC016467.1 | 10 | 1923 | AAH16467.1 | |
| BX537452.1 | 10 | 1943 | CAD97695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3174n100 | Chr.17:41778258 - 41843183 on Build GRCh38 | Gain |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map