Assay Details
Target Gene Details
Entrez Gene ID: | 8764 |
Gene Name: | TNF receptor superfamily member 14 |
Gene Aliases: |
ATAR, CD270, HVEA, HVEM, LIGHTR, TR2 |
Location: |
Chr.1:2555767-2565622 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF14 | NM_001297605.1 | NP_001284534.1 | ||
| NM_003820.3 | NP_003811.2 | |||
| XM_006711019.2 | XP_006711082.1 | |||
| XM_011542383.2 | XP_011540685.1 | |||
| XM_017002718.1 | XP_016858207.1 | |||
| XM_017002719.1 | XP_016858208.1 | |||
| XM_017002720.1 | XP_016858209.1 | |||
| XM_017002721.1 | XP_016858210.1 | |||
| AB208808.1 | BAD92045.1 | |||
| AF153978.1 | AAF75588.1 | |||
| AF373877.1 | AAL47717.1 | |||
| AF373878.1 | AAL47718.1 | |||
| AK124010.1 | ||||
| AK310076.1 | ||||
| AY358879.1 | AAQ89238.1 | |||
| BC002794.2 | AAH02794.1 | |||
| BC029848.1 | AAH29848.1 | |||
| BX343044.2 | ||||
| CR456909.1 | CAG33190.1 | |||
| U70321.1 | AAB58354.1 | |||
| U81232.1 | AAD00505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951557 | Chr.1:2266062 - 2564661 on Build GRCh38 | Deletion |
|
| nsv545077 | Chr.1:2428542 - 2622185 on Build GRCh38 | Gain |
|
| nsv545082 | Chr.1:2536231 - 2597243 on Build GRCh38 | Loss |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map