Assay Details
Target Gene Details
Entrez Gene ID: | 59284 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 7 |
Gene Aliases: |
- |
Location: |
Chr.19:53909425-53943944 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG7 | NM_031896.4 | 5 | 1386 | NP_114102.2 |
| XM_005259124.3 | 5 | 1267 | XP_005259181.1 | |
| XM_017027093.1 | 6 | 1413 | XP_016882582.1 | |
| AL537399.3 | 1 | 571 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064558 | Chr.19:53773468 - 53957491 on Build GRCh38 | Gain |
|
| nsv2541 | Chr.19:53901764 - 53946589 on Build GRCh38 | Deletion |
|
| nsv1063127 | Chr.19:53926876 - 54068977 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv833876 | Chr.19:53845956 - 53995117 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113745456] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map