Assay Details
Target Gene Details
Entrez Gene ID: | 6868 |
Gene Name: | ADAM metallopeptidase domain 17 |
Gene Aliases: |
ADAM18, CD156B, CSVP, NISBD, NISBD1, TACE |
Location: |
Chr.2:9488486-9555820 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM17 | NM_003183.5 | 19 | 3359 | NP_003174.3 |
| XM_011510375.2 | 19 | 3378 | XP_011508677.1 | |
| XM_011510376.2 | 19 | 3347 | XP_011508678.1 | |
| XM_011510377.2 | 14 | 2732 | XP_011508679.1 | |
| XM_017004785.1 | 19 | 3380 | XP_016860274.1 | |
| XM_017004786.1 | 18 | 3072 | XP_016860275.1 | |
| AL043281.2 | 1 | 182 | ||
| BC136783.1 | 19 | 3221 | ||
| BM725368.1 | 1 | 254 | ||
| BQ186514.1 | 1 | 252 | ||
| GQ402545.1 | 19 | 3327 | ||
| U92649.1 | 19 | 3308 | AAC39721.1 |
Target Gene Details
Entrez Gene ID: | 285148 |
Gene Name: | isoamyl acetate-hydrolyzing esterase 1 homolog |
Gene Aliases: |
- |
Location: |
Chr.2:9473638-9512403 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IAH1 | AK297648.1 | |||
| BC059410.1 | 5 | 1886 | AAH59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map