Assay Details
Target Gene Details
Entrez Gene ID: | 3762 |
Gene Name: | potassium voltage-gated channel subfamily J member 5 |
Gene Aliases: |
CIR, GIRK4, KATP1, KIR3.4, LQT13 |
Location: |
Chr.11:128891418-128921163 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ5 | NM_000890.3 | 1 | 119 | NP_000881.3 |
| AK312837.1 | 1 | 181 | ||
| BC069386.1 | 1 | 71 | AAH69386.1 | |
| BC069482.1 | 1 | 71 | AAH69482.1 | |
| BC069499.1 | 1 | 71 | AAH69499.1 | |
| BC069571.1 | 1 | 68 | AAH69571.1 | |
| D50134.1 | 1 | 75 | BAA08814.1 | |
| U39195.1 | AAB53093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628051 | Chr.11:128890799 - 128892879 on Build GRCh38 | Loss |
|
| esv2656616 | Chr.11:128890803 - 128892940 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map