Assay Details
Target Gene Details
Entrez Gene ID: | 92597 |
Gene Name: | MOB kinase activator 1B |
Gene Aliases: |
MATS2, MOB4A, MOBKL1A |
Location: |
Chr.4:70901970-70988174 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB1B | NM_001244766.1 | 1 | 60 | NP_001231695.1 |
| NM_001244767.1 | 1 | 60 | NP_001231696.1 | |
| NM_173468.3 | 1 | 60 | NP_775739.1 | |
| XM_005265709.2 | 1 | 69 | XP_005265766.1 | |
| XM_011532412.2 | XP_011530714.1 | |||
| AK093545.1 | 1 | 53 | ||
| AK299481.1 | ||||
| AK313513.1 | ||||
| BC038112.1 | AAH38112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594651 | Chr.4:70815237 - 70911218 on Build GRCh38 | Loss |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
| nsv829966 | Chr.4:70794825 - 70949164 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113511150] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map