Assay Details
Target Gene Details
Entrez Gene ID: | 79811 |
Gene Name: | SAFB like transcription modulator |
Gene Aliases: |
Met |
Location: |
Chr.15:58879045-58934313 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| nsv1035640 | Chr.15:58813161 - 58898731 on Build GRCh38 | Gain |
|
| esv34110 | Chr.15:58657792 - 59089435 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map