Assay Details
Target Gene Details
Entrez Gene ID: | 377677 |
Gene Name: | carbonic anhydrase 13 |
Gene Aliases: |
CAXIII |
Location: |
Chr.8:85245487-85284073 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA13 | NM_198584.2 | 1 | 309 | NP_940986.1 |
| AK095314.1 | BAC04528.1 | |||
| BC052602.1 | 1 | 269 | AAH52602.1 | |
| DB287770.1 | 1 | 309 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761441 | Chr.8:85243363 - 85254900 on Build GRCh38 | Loss |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map