Assay Details
Target Gene Details
Entrez Gene ID: | 4900 |
Gene Name: | neurogranin |
Gene Aliases: |
RC3, hng |
Location: |
Chr.11:124739933-124747206 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRGN | NM_001126181.1 | NP_001119653.1 | ||
| NM_006176.2 | NP_006167.1 | |||
| BC002835.2 | AAH02835.1 | |||
| U89165.1 | AAB49458.1 | |||
| Y09689.1 | CAA70867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627986 | Chr.11:124740300 - 124752449 on Build GRCh38 | Gain |
|
| nsv952751 | Chr.11:124744805 - 124748804 on Build GRCh38 | Deletion |
|
| nsv556487 | Chr.11:124730862 - 124765248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map