Assay Details
Target Gene Details
Entrez Gene ID: | 85397 |
Gene Name: | regulator of G-protein signaling 8 |
Gene Aliases: |
- |
Location: |
Chr.1:182641814-182753291 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS8 | NM_001102450.2 | 7 | 888 | NP_001095920.1 |
| NM_033345.3 | 6 | 759 | NP_203131.1 | |
| XM_005245555.3 | 7 | 902 | XP_005245612.1 | |
| XM_011510089.2 | 7 | 1799 | XP_011508391.1 | |
| XM_011510090.2 | 10 | 1629 | XP_011508392.1 | |
| XM_011510091.2 | 6 | 776 | XP_011508393.1 | |
| XM_017002631.1 | 10 | 1249 | XP_016858120.1 | |
| XM_017002632.1 | 8 | 1080 | XP_016858121.1 | |
| XM_017002633.1 | 7 | 1278 | XP_016858122.1 | |
| XM_017002634.1 | 7 | 957 | XP_016858123.1 | |
| XM_017002635.1 | 8 | 1404 | XP_016858124.1 | |
| XM_017002636.1 | 6 | 886 | XP_016858125.1 | |
| XM_017002637.1 | 7 | 1084 | XP_016858126.1 | |
| AI209102.1 | ||||
| AK294660.1 | 7 | 855 | ||
| AK296514.1 | 7 | 848 | ||
| BC069677.1 | AAH69677.1 | |||
| BC069718.1 | AAH69718.1 | |||
| BC103865.1 | AAI03866.1 | |||
| BC103866.1 | AAI03867.1 | |||
| BC103867.1 | AAI03868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3810 | Chr.1:182630610 - 182661590 on Build GRCh38 | Deletion |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map