Assay Details
Target Gene Details
Entrez Gene ID: | 51704 |
Gene Name: | G protein-coupled receptor class C group 5 member B |
Gene Aliases: |
RAIG-2, RAIG2 |
Location: |
Chr.16:19858971-19886045 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC5B | NM_001304771.1 | 2 | 989 | NP_001291700.1 |
| NM_016235.2 | 2 | 742 | NP_057319.1 | |
| XM_006721051.2 | 2 | 727 | XP_006721114.1 | |
| XM_006721052.2 | 2 | 677 | XP_006721115.1 | |
| AF181862.1 | 2 | 664 | AAF05331.1 | |
| AF202640.1 | 2 | 623 | AAF67321.1 | |
| AJ276101.1 | 2 | 671 | CAC00632.1 | |
| AK075119.1 | 2 | 674 | BAC11414.1 | |
| AK294680.1 | 3 | 382 | ||
| AK295204.1 | ||||
| AK296754.1 | ||||
| AK302838.1 | 2 | 865 | ||
| AK315989.1 | 3 | 406 | ||
| BC034467.2 | 2 | 655 | AAH34467.1 | |
| FJ529380.1 | 1 | 556 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428321 | Chr.16:19742033 - 19876673 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map