Assay Details
Target Gene Details
Entrez Gene ID: | 221955 |
Gene Name: | diacylglycerol lipase beta |
Gene Aliases: |
DAGLBETA, KCCR13L |
Location: |
Chr.7:6409116-6448012 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAGLB | NM_001142936.1 | 13 | 2105 | NP_001136408.1 |
| NM_139179.3 | 15 | 2492 | NP_631918.3 | |
| AF450090.1 | 15 | 2434 | AAL47020.1 | |
| AK074210.1 | 9 | 1374 | BAB85017.1 | |
| AK074584.1 | 6 | 1138 | BAC11073.1 | |
| AK074744.1 | 15 | 2432 | BAC11175.1 | |
| AK075128.1 | 15 | 2431 | BAC11420.1 | |
| AK090943.1 | 6 | 1973 | ||
| AK091228.1 | 7 | 2181 | ||
| AK093958.1 | 12 | 1829 | BAC04258.1 | |
| AK122748.1 | 12 | 1858 | ||
| AK298910.1 | 12 | 1818 | ||
| AK298955.1 | ||||
| BC027603.1 | 15 | 2371 | AAH27603.1 | |
| CB528359.1 | 1 | 427 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020236 | Chr.7:6209299 - 6650746 on Build GRCh38 | Gain |
|
| nsv606000 | Chr.7:6406396 - 6437371 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv605999 | Chr.7:6357088 - 6433250 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73341299] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map