Assay Details
Target Gene Details
Entrez Gene ID: | 9762 |
Gene Name: | leucine zipper tumor suppressor family member 3 |
Gene Aliases: |
PROSAPIP1 |
Location: |
Chr.20:3162617-3173597 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LZTS3 | NM_001282533.1 | 4 | 1239 | NP_001269462.1 |
| XM_005260890.3 | 4 | 1183 | XP_005260947.1 | |
| XM_005260892.1 | 4 | 1239 | XP_005260949.1 | |
| XM_005260893.2 | 4 | 1152 | XP_005260950.1 | |
| XM_011529407.2 | 4 | 1183 | XP_011527709.1 | |
| XM_011529408.2 | 4 | 2221 | XP_011527710.1 | |
| XM_011529409.2 | 4 | 2641 | XP_011527711.1 | |
| AB011124.1 | 2 | 2267 | BAA25478.2 | |
| BC038860.1 | 4 | 1195 | AAH38860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
| nsv833898 | Chr.20:3147423 - 3309891 on Build GRCh38 | Gain+Loss |
|

Genomic Map