Assay Details
Target Gene Details
Entrez Gene ID: | 27109 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B) |
Gene Aliases: |
ATPW, FB, HSU79253 |
Location: |
Chr.14:50311752-50326715 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| nsv832793 | Chr.14:50294245 - 50470857 on Build GRCh38 | Loss |
|
| nsv1052059 | Chr.14:50291535 - 50364621 on Build GRCh38 | Loss |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map