Assay Details
Target Gene Details
Entrez Gene ID: | 56648 |
Gene Name: | eukaryotic translation initiation factor 5A2 |
Gene Aliases: |
EIF-5A2, eIF5AII |
Location: |
Chr.3:170888415-170908637 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF5A2 | NM_020390.5 | 5 | 2786 | NP_065123.1 |
| AY205260.1 | 5 | 2786 | AAO18678.1 | |
| AY205261.1 | 5 | 2786 | AAO18679.1 | |
| BC036072.1 | 5 | 2757 | AAH36072.1 |
More Information
Set Membership: |
|

Genomic Map