Assay Details
Target Gene Details
Entrez Gene ID: | 79056 |
Gene Name: | proline rich and Gla domain 4 |
Gene Aliases: |
PRGP4, TMG4 |
Location: |
Chr.11:32829756-32858123 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRRG4 | NM_024081.5 | 2 | 284 | NP_076986.1 |
| XM_006718313.3 | 2 | 463 | XP_006718376.3 | |
| XM_006718314.3 | 2 | 134 | XP_006718377.1 | |
| XM_017018275.1 | 2 | 444 | XP_016873764.1 | |
| AF326351.1 | 2 | 132 | AAK00956.1 | |
| BC010052.2 | 2 | 284 | AAH10052.1 | |
| BC063393.1 | 2 | 276 | AAH63393.1 | |
| KU178766.1 | ||||
| KU178767.1 | ||||
| KU178768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759815 | Chr.11:32693097 - 33028394 on Build GRCh38 | Gain |
|
| esv34742 | Chr.11:32774546 - 32982478 on Build GRCh38 | Gain |
|
| nsv825818 | Chr.11:32828358 - 32830619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map