Assay Details
Target Gene Details
Entrez Gene ID: | 4665 |
Gene Name: | NGFI-A binding protein 2 |
Gene Aliases: |
MADER |
Location: |
Chr.12:57088894-57095476 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAB2 | NM_005967.3 | 6 | 1731 | NP_005958.1 |
| XM_005268894.2 | XP_005268951.1 | |||
| AK314229.1 | 6 | 1511 | ||
| BC065931.1 | 6 | 1518 | AAH65931.1 | |
| U48361.1 | 6 | 1510 | AAC50589.1 | |
| X70991.1 | 5 | 1203 | CAA50318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv832428 | Chr.12:56946825 - 57141911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map